Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTGGAAGTCACCTCTGTGAAT[A/G]AGCCTCTCCACGCAGAATGTGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608246 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT72 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT72 - keratin 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146225.1 | Intron | NP_001139697.1 | ||||
NM_001146226.1 | Intron | NP_001139698.1 | ||||
NM_080747.2 | Intron | NP_542785.1 | ||||
XM_017018833.1 | Intron | XP_016874322.1 | ||||
XM_017018834.1 | Intron | XP_016874323.1 | ||||
XM_017018835.1 | Intron | XP_016874324.1 | ||||
XM_017018836.1 | Intron | XP_016874325.1 | ||||
XM_017018837.1 | Intron | XP_016874326.1 |