Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCAGAATGCTGGTGCCAGTGGC[C/T]GCCACCTCCTTCCCAGGTTTTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf89 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf89 - chromosome 17 open reading frame 89 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC38A10 - solute carrier family 38 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037984.2 | 2512 | Silent Mutation | GCC,GCT | A,A 900 | NP_001033073.1 | |
NM_138570.3 | 2512 | Intron | NP_612637.1 | |||
XM_005257019.1 | 2512 | Silent Mutation | GCC,GCT | A,A 908 | XP_005257076.1 | |
XM_011524288.1 | 2512 | Silent Mutation | GCC,GCT | A,A 915 | XP_011522590.1 | |
XM_011524289.1 | 2512 | Silent Mutation | GCC,GCT | A,A 907 | XP_011522591.1 | |
XM_011524290.1 | 2512 | Silent Mutation | GCC,GCT | A,A 826 | XP_011522592.1 |
TEPSIN - TEPSIN, adaptor related protein complex 4 accessory protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |