Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGCAGGGATGGGACTCAGGAAAG[A/G]GAAGCATGTGTGTGTTGAATAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNASE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNASE1 - ribonuclease A family member 1, pancreatic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002933.4 | 776 | UTR 3 | NP_002924.1 | |||
NM_198232.2 | 776 | UTR 3 | NP_937875.1 | |||
NM_198234.2 | 776 | UTR 3 | NP_937877.1 | |||
NM_198235.2 | 776 | UTR 3 | NP_937878.1 |