Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCTGCACGTCATTGAGGACATT[C/T]TTACTATCTCCAATGTAGTCTAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610096 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIMD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TIMD4 - T-cell immunoglobulin and mucin domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146726.1 | 1050 | Silent Mutation | AAA,AAG | K,K 331 | NP_001140198.1 | |
NM_138379.2 | 1050 | Silent Mutation | AAA,AAG | K,K 359 | NP_612388.2 | |
XM_011534694.2 | 1050 | Intron | XP_011532996.1 | |||
XM_017010021.1 | 1050 | Silent Mutation | AAA,AAG | K,K 304 | XP_016865510.1 |