Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGACGTAGGATCGCGCCCAGGAG[A/G]GGGCGCTGTGCTGGGTCAGGCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615792 MIM: 603921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT15 - nudix hydrolase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304745.1 | Intron | NP_001291674.1 | ||||
NM_018283.3 | Intron | NP_060753.1 |
SUCLA2 - succinate-CoA ligase ADP-forming beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003850.2 | Intron | NP_003841.1 | ||||
XM_017020806.1 | Intron | XP_016876295.1 | ||||
XM_017020807.1 | Intron | XP_016876296.1 | ||||
XM_017020808.1 | Intron | XP_016876297.1 | ||||
XM_017020809.1 | Intron | XP_016876298.1 |