Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTTCTGGACTACAAACAAACC[A/G]TTCACAATTGTTCCAAGGGGCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604578 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIM2 - absent in melanoma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004833.1 | 1040 | Silent Mutation | AAC,AAT | N,N 265 | NP_004824.1 | |
XM_005245616.4 | 1040 | Silent Mutation | AAC,AAT | N,N 160 | XP_005245673.1 | |
XM_017002848.1 | 1040 | Silent Mutation | AAC,AAT | N,N 265 | XP_016858337.1 |