Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACGAGGGGGCAACAGCTGGGGG[A/C]GGCTTCTTCAGGGGGTTGCCATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605639 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIGLEC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIGLEC8 - sialic acid binding Ig like lectin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014442.2 | 1321 | Silent Mutation | CCG,CCT | P,P 431 | NP_055257.2 | |
XM_011526734.2 | 1321 | Silent Mutation | CCG,CCT | P,P 420 | XP_011525036.1 | |
XM_011526735.2 | 1321 | Silent Mutation | CCG,CCT | P,P 338 | XP_011525037.1 |