Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTCGGGGGCCAAGTCCGGCAGC[A/G]GTACCTCTACACAGATGATGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609436 MIM: 211100 MIM: 609278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF21 - fibroblast growth factor 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019113.3 | 712 | Missense Mutation | CAG,CGG | Q,R 47 | NP_061986.1 |
FUT1 - fucosyltransferase 1 (H blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000148.3 | 712 | Intron | NP_000139.1 | |||
XM_006723127.2 | 712 | Intron | XP_006723190.1 | |||
XM_017026552.1 | 712 | Intron | XP_016882041.1 | |||
XM_017026553.1 | 712 | Intron | XP_016882042.1 |
IZUMO1 - izumo sperm-egg fusion 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |