Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGCCGTGTTCTCCTGCTCGAC[A/G]TTGATCCACTCTCTTCTCTTTTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146790 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGR2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FCGR2A - Fc fragment of IgG receptor IIa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136219.1 | Intron | NP_001129691.1 | ||||
NM_021642.3 | Intron | NP_067674.2 | ||||
XM_011509287.2 | Intron | XP_011507589.1 | ||||
XM_011509290.2 | Intron | XP_011507592.1 | ||||
XM_011509291.1 | Intron | XP_011507593.1 | ||||
XM_017000663.1 | Intron | XP_016856152.1 | ||||
XM_017000664.1 | Intron | XP_016856153.1 | ||||
XM_017000665.1 | Intron | XP_016856154.1 | ||||
XM_017000666.1 | Intron | XP_016856155.1 | ||||
XM_017000667.1 | Intron | XP_016856156.1 | ||||
XM_017000668.1 | Intron | XP_016856157.1 |
Set Membership: |
HapMap |