Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGACCGGCCAGGAGCTCTGCCC[C/T]CAGGGCTCCTATTTGAGGATGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSBPL5 - oxysterol binding protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144063.1 | 3154 | UTR 3 | NP_001137535.1 | |||
NM_020896.3 | 3154 | UTR 3 | NP_065947.1 | |||
NM_145638.2 | 3154 | UTR 3 | NP_663613.1 | |||
XM_011519873.2 | 3154 | UTR 3 | XP_011518175.1 | |||
XM_017017163.1 | 3154 | UTR 3 | XP_016872652.1 | |||
XM_017017164.1 | 3154 | UTR 3 | XP_016872653.1 |