Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTAGCGCACAGAGGACAATCCT[C/T]ACCAACACGCCACCAACCACTCCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190197 MIM: 600697 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CNTN2 - contactin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBBP5 - RB binding protein 5, histone lysine methyltransferase complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM81 - transmembrane protein 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203376.1 | 866 | Silent Mutation | GTA,GTG | V,V 242 | NP_976310.1 |