Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTATGTGCACTTTGGTCTTGTGTA[A/G]TGTTCCTTTAATGAATCATTTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V0A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V0A1 - ATPase H+ transporting V0 subunit a1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130020.1 | Intron | NP_001123492.1 | ||||
NM_001130021.1 | Intron | NP_001123493.1 | ||||
NM_005177.3 | Intron | NP_005168.2 | ||||
XM_005257459.1 | Intron | XP_005257516.1 | ||||
XM_005257461.1 | Intron | XP_005257518.1 | ||||
XM_005257463.2 | Intron | XP_005257520.1 | ||||
XM_011524908.1 | Intron | XP_011523210.1 | ||||
XM_017024766.1 | Intron | XP_016880255.1 | ||||
XM_017024767.1 | Intron | XP_016880256.1 | ||||
XM_017024768.1 | Intron | XP_016880257.1 | ||||
XM_017024769.1 | Intron | XP_016880258.1 | ||||
XM_017024770.1 | Intron | XP_016880259.1 | ||||
XM_017024771.1 | Intron | XP_016880260.1 | ||||
XM_017024772.1 | Intron | XP_016880261.1 | ||||
XM_017024773.1 | Intron | XP_016880262.1 | ||||
XM_017024774.1 | Intron | XP_016880263.1 |