Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACTGTTATTATGTATTTTTGTG[A/T]AAATGCAATAATTATTTGACCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603317 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRQ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRQ - protein tyrosine phosphatase, receptor type Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145026.1 | Intron | NP_001138498.1 | ||||
XM_017019273.1 | Intron | XP_016874762.1 | ||||
XM_017019274.1 | Intron | XP_016874763.1 | ||||
XM_017019275.1 | Intron | XP_016874764.1 |