Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACATATGTCCCCCGGCTGGTAA[A/G]GATGAAGGTGTTCCACCTCAGCCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613276 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USB1 - U6 snRNA biogenesis phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195302.1 | 421 | Missense Mutation | AAG,AGG | K,R 115 | NP_001182231.1 | |
NM_001204911.1 | 421 | Missense Mutation | AAG,AGG | K,R 115 | NP_001191840.1 | |
NM_024598.3 | 421 | Missense Mutation | AAG,AGG | K,R 115 | NP_078874.2 | |
XM_005256144.3 | 421 | Missense Mutation | AAG,AGG | K,R 64 | XP_005256201.1 | |
XM_011523328.2 | 421 | Missense Mutation | AAG,AGG | K,R 102 | XP_011521630.1 | |
XM_011523329.2 | 421 | Missense Mutation | AAG,AGG | K,R 64 | XP_011521631.1 | |
XM_011523330.1 | 421 | Missense Mutation | AAG,AGG | K,R 115 | XP_011521632.1 |
ZNF319 - zinc finger protein 319 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |