Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGCCATGGCACCTTGGCTGAGA[G/T]AACGGCCCGCCCTCTTTACTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USB1 - U6 snRNA biogenesis phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195302.1 | Intron | NP_001182231.1 | ||||
NM_001204911.1 | Intron | NP_001191840.1 | ||||
NM_024598.3 | Intron | NP_078874.2 | ||||
XM_005256144.3 | Intron | XP_005256201.1 | ||||
XM_011523328.2 | Intron | XP_011521630.1 | ||||
XM_011523329.2 | Intron | XP_011521631.1 | ||||
XM_011523330.1 | Intron | XP_011521632.1 |
ZNF319 - zinc finger protein 319 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |