Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATTAAATGTCAGCTATTTAGGG[C/T]CTGCTTCATTTATATTAAATAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612723 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLEKHH2 - pleckstrin homology, MyTH4 and FERM domain containing H2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172069.3 | Intron | NP_742066.2 | ||||
XM_017003351.1 | Intron | XP_016858840.1 | ||||
XM_017003352.1 | Intron | XP_016858841.1 | ||||
XM_017003353.1 | Intron | XP_016858842.1 |