Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAGACTCTTACGAACAAGGTGA[C/T]TCTTCCCAGCAGTCCTTGAAGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACYP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACYP1 - acylphosphatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZC2HC1C - zinc finger C2HC-type containing 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042430.2 | 245 | Silent Mutation | GAC,GAT | D,D 39 | NP_001035895.1 | |
NM_024643.3 | 245 | Silent Mutation | GAC,GAT | D,D 39 | NP_078919.2 | |
XM_005268062.3 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_005268119.1 | |
XM_005268063.4 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_005268120.1 | |
XM_006720255.3 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_006720318.1 | |
XM_011537155.2 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_011535457.1 | |
XM_011537156.2 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_011535458.1 | |
XM_011537157.2 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_011535459.1 | |
XM_017021643.1 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_016877132.1 | |
XM_017021644.1 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_016877133.1 | |
XM_017021645.1 | 245 | Silent Mutation | GAC,GAT | D,D 39 | XP_016877134.1 |