Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAATTTTGAGATCACTGAGAAGC[A/C]AAAACAGTCTGTTACGTGTTCATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616173 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB5 - EF-hand calcium binding domain 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3184 - microRNA 3184 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR423 - microRNA 423 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSRP1 - nuclear speckle splicing regulatory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261467.1 | Intron | NP_001248396.1 | ||||
NM_032141.3 | Intron | NP_115517.1 | ||||
XM_011525345.2 | Intron | XP_011523647.1 | ||||
XM_017025209.1 | Intron | XP_016880698.1 | ||||
XM_017025210.1 | Intron | XP_016880699.1 | ||||
XM_017025211.1 | Intron | XP_016880700.1 | ||||
XM_017025212.1 | Intron | XP_016880701.1 | ||||
XM_017025213.1 | Intron | XP_016880702.1 | ||||
XM_017025214.1 | Intron | XP_016880703.1 | ||||
XM_017025215.1 | Intron | XP_016880704.1 |