Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGACATGGTGGGGCTCCAGGC[A/G]GAGCAGGGCGAGCATGGCAGTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRPF4 - pre-mRNA processing factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF183 - ring finger protein 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145051.3 | 1964 | Missense Mutation | CGC,TGC | R,C 85 | NP_659488.2 | |
XM_006716952.2 | 1964 | Missense Mutation | CGC,TGC | R,C 85 | XP_006717015.1 | |
XM_011518218.1 | 1964 | Missense Mutation | CGC,TGC | R,C 85 | XP_011516520.1 | |
XM_011518219.1 | 1964 | Missense Mutation | CGC,TGC | R,C 85 | XP_011516521.1 | |
XM_011518220.1 | 1964 | Missense Mutation | CGC,TGC | R,C 85 | XP_011516522.1 | |
XM_017014268.1 | 1964 | Missense Mutation | CGC,TGC | R,C 85 | XP_016869757.1 |