Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAAGCAGGAAGAGAGATGCAGT[A/G]TTGGGCATTCAGTCAGGCCCCTGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608107 MIM: 603231 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEFV PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MEFV - Mediterranean fever | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000243.2 | 2533 | UTR 3 | NP_000234.1 | |||
NM_001198536.1 | 2533 | UTR 3 | NP_001185465.1 | |||
XM_017023236.1 | 2533 | UTR 3 | XP_016878725.1 |
ZNF200 - zinc finger protein 200 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |