Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147045 MIM: 604947 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCAR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCAR - Fc fragment of IgA receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIR3DL2 - killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242867.1 | 1242 | Intron | NP_001229796.1 | |||
NM_006737.3 | 1242 | Intron | NP_006728.2 | |||
XM_011526940.2 | 1242 | Missense Mutation | ACG,ATG | T,M 362 | XP_011525242.1 | |
XM_017026784.1 | 1242 | Intron | XP_016882273.1 |
Set Membership: |
HapMap |