Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCCATTTTATTTCCTCAGAATG[A/G]ACTCAGGGAAGTGAGAGAAGAGCTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX35 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TEX35 - testis expressed 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170722.1 | 276 | Missense Mutation | GAA,GGA | E,G 63 | NP_001164193.1 | |
NM_001170723.1 | 276 | Missense Mutation | GAA,GGA | E,G 55 | NP_001164194.1 | |
NM_001170724.1 | 276 | Missense Mutation | GAA,GGA | E,G 55 | NP_001164195.1 | |
NM_032126.4 | 276 | Missense Mutation | GAA,GGA | E,G 55 | NP_115502.2 |