Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCTTTTTACTGGTCTCACCCA[A/G]AATTCATGCAGAGGTTTTGTTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613833 MIM: 180480 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANSL1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KANSL1L - KAT8 regulatory NSL complex subunit 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307976.1 | 5975 | Intron | NP_001294905.1 | |||
NM_152519.3 | 5975 | Intron | NP_689732.2 | |||
XM_005246328.4 | 5975 | Intron | XP_005246385.1 | |||
XM_005246329.4 | 5975 | Intron | XP_005246386.1 | |||
XM_005246330.4 | 5975 | Intron | XP_005246387.1 | |||
XM_005246332.4 | 5975 | Intron | XP_005246389.1 | |||
XM_005246334.4 | 5975 | Intron | XP_005246391.1 | |||
XM_006712320.3 | 5975 | Intron | XP_006712383.1 | |||
XM_011510706.2 | 5975 | Intron | XP_011509008.1 | |||
XM_011510707.2 | 5975 | Intron | XP_011509009.1 | |||
XM_011510709.2 | 5975 | Intron | XP_011509011.1 | |||
XM_011510710.2 | 5975 | Intron | XP_011509012.1 | |||
XM_017003430.1 | 5975 | Intron | XP_016858919.1 | |||
XM_017003431.1 | 5975 | Intron | XP_016858920.1 |
LOC101928020 - uncharacterized LOC101928020 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPE - ribulose-5-phosphate-3-epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278282.1 | 5975 | Intron | NP_001265211.1 | |||
NM_001278283.1 | 5975 | Intron | NP_001265212.1 | |||
NM_001278285.1 | 5975 | Intron | NP_001265214.1 | |||
NM_001278286.1 | 5975 | Intron | NP_001265215.1 | |||
NM_001278288.1 | 5975 | Intron | NP_001265217.1 | |||
NM_001278289.1 | 5975 | Intron | NP_001265218.1 | |||
NM_001318926.1 | 5975 | Intron | NP_001305855.1 | |||
NM_001318927.1 | 5975 | Intron | NP_001305856.1 | |||
NM_001318928.1 | 5975 | Intron | NP_001305857.1 | |||
NM_001318929.1 | 5975 | Intron | NP_001305858.1 | |||
NM_001318930.1 | 5975 | Intron | NP_001305859.1 | |||
NM_001318931.1 | 5975 | Intron | NP_001305860.1 | |||
NM_006916.2 | 5975 | Intron | NP_008847.1 | |||
NM_199229.2 | 5975 | Intron | NP_954699.1 | |||
XM_006712677.3 | 5975 | UTR 3 | XP_006712740.1 |