Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601232 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3CG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PIK3CG - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282426.1 | 194 | Silent Mutation | TGC,TGT | C,C 17 | NP_001269355.1 | |
NM_001282427.1 | 194 | Silent Mutation | TGC,TGT | C,C 17 | NP_001269356.1 | |
NM_002649.3 | 194 | Silent Mutation | TGC,TGT | C,C 17 | NP_002640.2 | |
XM_005250443.3 | 194 | Silent Mutation | TGC,TGT | C,C 17 | XP_005250500.1 | |
XM_011516316.1 | 194 | Silent Mutation | TGC,TGT | C,C 17 | XP_011514618.1 | |
XM_011516317.1 | 194 | Silent Mutation | TGC,TGT | C,C 17 | XP_011514619.1 | |
XM_017012328.1 | 194 | Silent Mutation | TGC,TGT | C,C 17 | XP_016867817.1 |
Set Membership: |
HapMap |