Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCGAGTATTCATCAACTGGTCTC[A/C]ATTTCCTGAACACATTCACTGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615936 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP42 - Rho GTPase activating protein 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152432.2 | 509 | Intron | NP_689645.2 | |||
XM_011542615.2 | 509 | UTR 3 | XP_011540917.1 | |||
XM_011542616.2 | 509 | UTR 3 | XP_011540918.1 | |||
XM_011542617.2 | 509 | UTR 3 | XP_011540919.1 | |||
XM_017017237.1 | 509 | UTR 3 | XP_016872726.1 | |||
XM_017017238.1 | 509 | UTR 3 | XP_016872727.1 |
TMEM133 - transmembrane protein 133 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032021.2 | 509 | Missense Mutation | AAT,CAT | N,H 94 | NP_114410.1 |