Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGAATCTGGTTTTTGAGGACCA[A/T]TTTCTGCCTCCTGGCATCTTCCATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607334 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRAK2 - trafficking kinesin protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015049.2 | 2938 | Missense Mutation | AAT,ATT | N,I 863 | NP_055864.2 | |
XM_011511690.1 | 2938 | Missense Mutation | AAT,ATT | N,I 863 | XP_011509992.1 | |
XM_017004772.1 | 2938 | Missense Mutation | AAT,ATT | N,I 652 | XP_016860261.1 |
Set Membership: |
HapMap |