Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATCTGCGACGCTTAGCAGCCC[C/T]GTTCCAGCCTCATCATAGCTTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB6 - EF-hand calcium binding domain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022785.3 | 4365 | Silent Mutation | ACA,ACG | T,T 1451 | NP_073622.2 | |
NM_198856.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1299 | NP_942153.1 | |
XM_005261704.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1451 | XP_005261761.1 | |
XM_005261705.3 | 4365 | Silent Mutation | ACA,ACG | T,T 1345 | XP_005261762.1 | |
XM_011530316.1 | 4365 | Silent Mutation | ACA,ACG | T,T 1477 | XP_011528618.1 | |
XM_011530317.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1451 | XP_011528619.1 | |
XM_011530318.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1425 | XP_011528620.1 | |
XM_011530319.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1345 | XP_011528621.1 | |
XM_011530320.2 | 4365 | Missense Mutation | CAG,CGG | Q,R 1476 | XP_011528622.1 | |
XM_011530321.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1299 | XP_011528623.1 | |
XM_011530322.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1299 | XP_011528624.1 | |
XM_011530323.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1297 | XP_011528625.1 | |
XM_011530325.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1203 | XP_011528627.1 | |
XM_011530326.2 | 4365 | Intron | XP_011528628.1 | |||
XM_011530327.2 | 4365 | Silent Mutation | ACA,ACG | T,T 1143 | XP_011528629.1 | |
XM_011530328.2 | 4365 | Intron | XP_011528630.1 | |||
XM_011530329.1 | 4365 | Silent Mutation | ACA,ACG | T,T 749 | XP_011528631.1 | |
XM_011530330.2 | 4365 | Intron | XP_011528632.1 | |||
XM_017028910.1 | 4365 | Silent Mutation | ACA,ACG | T,T 1477 | XP_016884399.1 | |
XM_017028911.1 | 4365 | Silent Mutation | ACA,ACG | T,T 1477 | XP_016884400.1 | |
XM_017028912.1 | 4365 | Silent Mutation | ACA,ACG | T,T 1293 | XP_016884401.1 |
EFCAB6-AS1 - EFCAB6 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |