Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGGTTTCAAATTCATTGGTAA[C/T]AAATATGCTAACTTTCTGAATCAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606585 MIM: 147790 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENAM PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ENAM - enamelin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JCHAIN - joining chain of multimeric IgA and IgM | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144646.3 | 598 | UTR 3 | NP_653247.1 | |||
XM_006714211.2 | 598 | UTR 3 | XP_006714274.1 | |||
XM_006714212.2 | 598 | UTR 3 | XP_006714275.1 | |||
XM_011531925.1 | 598 | UTR 3 | XP_011530227.1 | |||
XM_011531926.1 | 598 | UTR 3 | XP_011530228.1 |
Set Membership: |
HapMap Validated |