Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608121 MIM: 608122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARVB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARVB - parvin beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PARVG - parvin gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137605.2 | 78 | UTR 5 | NP_001131077.1 | |||
NM_022141.6 | 78 | Intron | NP_071424.1 | |||
XM_005261702.3 | 78 | Intron | XP_005261759.1 | |||
XM_011530302.2 | 78 | Missense Mutation | CGT,TGT | R,C 11 | XP_011528604.1 | |
XM_017028907.1 | 78 | Intron | XP_016884396.1 | |||
XM_017028908.1 | 78 | Intron | XP_016884397.1 |