Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAAAAGGTGATGAGGAGGAAAG[G/T]CTGTCACAACTCCTGGGAAGTATCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611359 MIM: 615086 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMBRA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMBRA1 - autophagy and beclin 1 regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HARBI1 - harbinger transposase derived 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173811.3 | 1699 | UTR 3 | NP_776172.1 | |||
XM_011520025.1 | 1699 | UTR 3 | XP_011518327.1 | |||
XM_011520026.2 | 1699 | UTR 3 | XP_011518328.1 | |||
XM_011520027.1 | 1699 | UTR 3 | XP_011518329.1 | |||
XM_011520028.2 | 1699 | UTR 3 | XP_011518330.1 | |||
XM_011520029.2 | 1699 | UTR 3 | XP_011518331.1 | |||
XM_017017602.1 | 1699 | UTR 3 | XP_016873091.1 | |||
XM_017017603.1 | 1699 | UTR 3 | XP_016873092.1 | |||
XM_017017604.1 | 1699 | UTR 3 | XP_016873093.1 | |||
XM_017017605.1 | 1699 | UTR 3 | XP_016873094.1 | |||
XM_017017606.1 | 1699 | UTR 3 | XP_016873095.1 |