Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACGGAGCGCGCAGCCCCGCGACA[C/T]CGTCATTGAAAAATTGGGGTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604586 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STXBP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STXBP5 - syntaxin binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127715.2 | Intron | NP_001121187.1 | ||||
NM_139244.4 | Intron | NP_640337.3 | ||||
XM_006715339.2 | Intron | XP_006715402.1 | ||||
XM_011535453.1 | Intron | XP_011533755.1 | ||||
XM_017010267.1 | Intron | XP_016865756.1 | ||||
XM_017010268.1 | Intron | XP_016865757.1 |
STXBP5-AS1 - STXBP5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |