Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTAAGTCAGGCCAGTGTCCTCCC[A/T]GCTGCAGAGCTGAGAAGGCTGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610106 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBNL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBNL - drebrin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014436.2 | Intron | NP_001014436.1 | ||||
NM_001122956.1 | Intron | NP_001116428.1 | ||||
NM_001284313.1 | Intron | NP_001271242.1 | ||||
NM_001284315.1 | Intron | NP_001271244.1 | ||||
NM_014063.6 | Intron | NP_054782.2 | ||||
XM_011515325.1 | Intron | XP_011513627.1 |
LINC00957 - long intergenic non-protein coding RNA 957 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6837 - microRNA 6837 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASA4CP - RAS p21 protein activator 4C, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |