Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300499 MIM: 300649 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FTSJ1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FTSJ1 - FtsJ RNA methyltransferase homolog 1 (E. coli) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282157.1 | Intron | NP_001269086.1 | ||||
NM_012280.3 | Intron | NP_036412.1 | ||||
NM_177439.2 | Intron | NP_803188.1 | ||||
XM_005272595.1 | Intron | XP_005272652.1 | ||||
XM_011543893.2 | Intron | XP_011542195.1 | ||||
XM_011543894.2 | Intron | XP_011542196.1 | ||||
XM_011543895.1 | Intron | XP_011542197.1 | ||||
XM_017029382.1 | Intron | XP_016884871.1 |
SLC38A5 - solute carrier family 38 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |