Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTATATGCAGGCCCAGCTCCTC[T/C]TAGCTGTGAGTAACCCTGGCCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300499 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FTSJ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FTSJ1 - FtsJ RNA methyltransferase homolog 1 (E. coli) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282157.1 | 827 | Intron | NP_001269086.1 | |||
NM_012280.3 | 827 | Silent Mutation | CTA,TTA | L,L 137 | NP_036412.1 | |
NM_177439.2 | 827 | Silent Mutation | CTA,TTA | L,L 137 | NP_803188.1 | |
XM_005272595.1 | 827 | Silent Mutation | CTA,TTA | L,L 137 | XP_005272652.1 | |
XM_011543893.2 | 827 | Silent Mutation | CTA,TTA | L,L 137 | XP_011542195.1 | |
XM_011543894.2 | 827 | Silent Mutation | CTA,TTA | L,L 137 | XP_011542196.1 | |
XM_011543895.1 | 827 | Silent Mutation | CTA,TTA | L,L 137 | XP_011542197.1 | |
XM_017029382.1 | 827 | Silent Mutation | CTA,TTA | L,L 137 | XP_016884871.1 |