Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCCGAGGCCGGGATGTGACGCT[T/C]CTCTACAGCCAGCTGCAGGTCTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300499 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FTSJ1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FTSJ1 - FtsJ RNA methyltransferase homolog 1 (E. coli) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282157.1 | 913 | Silent Mutation | CTC,CTT | L,L 28 | NP_001269086.1 | |
NM_012280.3 | 913 | Silent Mutation | CTC,CTT | L,L 165 | NP_036412.1 | |
NM_177439.2 | 913 | Silent Mutation | CTC,CTT | L,L 165 | NP_803188.1 | |
XM_005272595.1 | 913 | Silent Mutation | CTC,CTT | L,L 165 | XP_005272652.1 | |
XM_011543893.2 | 913 | Silent Mutation | CTC,CTT | L,L 165 | XP_011542195.1 | |
XM_011543894.2 | 913 | Silent Mutation | CTC,CTT | L,L 165 | XP_011542196.1 | |
XM_011543895.1 | 913 | Silent Mutation | CTC,CTT | L,L 165 | XP_011542197.1 | |
XM_017029382.1 | 913 | Silent Mutation | CTC,CTT | L,L 165 | XP_016884871.1 |
Set Membership: |
HapMap |