Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAAAGATGTACCTGGTTTGATC[A/G]TTTTTGCTTTTTCTGCTGGTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613323 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMPD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMPD2 - FERM and PDZ domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018071.3 | 2428 | Missense Mutation | NP_001018081.3 | |||
NM_001042512.2 | 2428 | Intron | NP_001035977.2 | |||
NM_001318191.1 | 2428 | Missense Mutation | NP_001305120.1 | |||
XM_011539327.2 | 2428 | Intron | XP_011537629.1 | |||
XM_017015744.1 | 2428 | Intron | XP_016871233.1 |