Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGCATTACCTCCAGCAACAGTC[G/A]ATAACAGGATTATTTTTGGAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605228 MIM: 614967 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CIR1 - corepressor interacting with RBPJ, 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRN3 - secernin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193528.1 | 1147 | Intron | NP_001180457.1 | |||
NM_024583.4 | 1147 | Missense Mutation | AAT,GAT | N,D 18 | NP_078859.2 | |
XM_005246853.2 | 1147 | Missense Mutation | AAT,GAT | N,D 38 | XP_005246910.1 | |
XM_005246854.4 | 1147 | Missense Mutation | AAT,GAT | N,D 18 | XP_005246911.1 | |
XM_005246855.2 | 1147 | Missense Mutation | AAT,GAT | N,D 18 | XP_005246912.1 | |
XM_005246856.2 | 1147 | Missense Mutation | AAT,GAT | N,D 18 | XP_005246913.1 | |
XM_005246857.2 | 1147 | Missense Mutation | AAT,GAT | N,D 18 | XP_005246914.1 | |
XM_011511839.2 | 1147 | Intron | XP_011510141.1 | |||
XM_017004909.1 | 1147 | Missense Mutation | AAT,GAT | N,D 18 | XP_016860398.1 | |
XM_017004910.1 | 1147 | Missense Mutation | AAT,GAT | N,D 18 | XP_016860399.1 |
Set Membership: |
HapMap |