Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGAAAAGGCCAGGTAAAATCA[C/T]CTTTGTATAGAGCTGAAGAATAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600655 MIM: 157655 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EEF1B2 - eukaryotic translation elongation factor 1 beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037663.1 | Intron | NP_001032752.1 | ||||
NM_001959.3 | Intron | NP_001950.1 | ||||
NM_021121.3 | Intron | NP_066944.1 |
NDUFS1 - NADH:ubiquinone oxidoreductase core subunit S1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA41 - small nucleolar RNA, H/ACA box 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD51 - small nucleolar RNA, C/D box 51 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |