Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN25 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZSCAN25 - zinc finger and SCAN domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145115.2 | Intron | NP_660090.2 | ||||
XM_005250194.2 | Intron | XP_005250251.1 | ||||
XM_011515905.2 | Intron | XP_011514207.1 | ||||
XM_011515906.2 | Intron | XP_011514208.1 | ||||
XM_011515907.2 | Intron | XP_011514209.1 | ||||
XM_011515908.2 | Intron | XP_011514210.1 | ||||
XM_011515909.2 | Intron | XP_011514211.1 | ||||
XM_011515910.2 | Intron | XP_011514212.1 | ||||
XM_017011824.1 | Intron | XP_016867313.1 | ||||
XM_017011825.1 | Intron | XP_016867314.1 | ||||
XM_017011826.1 | Intron | XP_016867315.1 | ||||
XM_017011827.1 | Intron | XP_016867316.1 |
Set Membership: |
HapMap Validated |