Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTGGGGTCCTGTAAGGGTCTAC[A/G]TGTGAAGAAGGATTTTTTACGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NNT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NNT - nicotinamide nucleotide transhydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012343.3 | 301 | Missense Mutation | CAT,CGT | H,R 27 | NP_036475.3 | |
NM_182977.2 | 301 | Missense Mutation | CAT,CGT | H,R 27 | NP_892022.2 | |
XM_005248274.4 | 301 | Missense Mutation | CAT,CGT | H,R 27 | XP_005248331.1 | |
XM_005248275.4 | 301 | Intron | XP_005248332.1 | |||
XM_006714461.3 | 301 | Intron | XP_006714524.1 | |||
XM_011514001.2 | 301 | Missense Mutation | CAT,CGT | H,R 27 | XP_011512303.1 | |
XM_011514002.2 | 301 | UTR 5 | XP_011512304.1 | |||
XM_017009293.1 | 301 | Missense Mutation | CAT,CGT | H,R 27 | XP_016864782.1 |
NNT-AS1 - NNT antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |