Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCCCCTTCAGTGCCATCACTC[A/G]TCGGCATCTGGCTTGACGTCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETS1 - ETS proto-oncogene 1, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143820.1 | 1311 | Silent Mutation | GAC,GAT | D,D 484 | NP_001137292.1 | |
NM_001162422.1 | 1311 | Silent Mutation | GAC,GAT | D,D 224 | NP_001155894.1 | |
NM_005238.3 | 1311 | Silent Mutation | GAC,GAT | D,D 440 | NP_005229.1 | |
XM_011542649.2 | 1311 | Silent Mutation | GAC,GAT | D,D 518 | XP_011540951.1 | |
XM_011542650.1 | 1311 | Silent Mutation | GAC,GAT | D,D 474 | XP_011540952.1 | |
XM_011542651.1 | 1311 | Silent Mutation | GAC,GAT | D,D 431 | XP_011540953.1 | |
XM_011542652.1 | 1311 | Silent Mutation | GAC,GAT | D,D 353 | XP_011540954.1 | |
XM_017017314.1 | 1311 | Silent Mutation | GAC,GAT | D,D 484 | XP_016872803.1 | |
XM_017017315.1 | 1311 | Silent Mutation | GAC,GAT | D,D 449 | XP_016872804.1 | |
XM_017017316.1 | 1311 | Silent Mutation | GAC,GAT | D,D 417 | XP_016872805.1 | |
XM_017017317.1 | 1311 | Silent Mutation | GAC,GAT | D,D 397 | XP_016872806.1 |