Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACAGGTTCCTTTCTATCTACCTC[A/T]ATTTTAGCTTTTCTACATTCCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606704 MIM: 607705 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR75 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR75 - G protein-coupled receptor 75 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR75-ASB3 - GPR75-ASB3 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSME4 - proteasome activator subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014614.2 | Intron | NP_055429.2 | ||||
XM_006711969.1 | Intron | XP_006712032.1 | ||||
XM_011532705.2 | Intron | XP_011531007.1 | ||||
XM_011532706.2 | Intron | XP_011531008.1 | ||||
XM_011532707.2 | Intron | XP_011531009.1 | ||||
XM_011532709.2 | Intron | XP_011531011.1 |