Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194526 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF34 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF34 - zinc finger protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286769.1 | 894 | Silent Mutation | AGA,AGG | R,R 240 | NP_001273698.1 | |
NM_001286770.1 | 894 | Silent Mutation | AGA,AGG | R,R 221 | NP_001273699.1 | |
NM_030580.4 | 894 | Silent Mutation | AGA,AGG | R,R 261 | NP_085057.3 | |
XM_011517314.2 | 894 | Silent Mutation | AGA,AGG | R,R 261 | XP_011515616.1 | |
XM_011517315.2 | 894 | Silent Mutation | AGA,AGG | R,R 240 | XP_011515617.1 | |
XM_011517316.2 | 894 | Silent Mutation | AGA,AGG | R,R 240 | XP_011515618.1 | |
XM_011517318.2 | 894 | Silent Mutation | AGA,AGG | R,R 240 | XP_011515620.1 | |
XM_011517320.2 | 894 | Silent Mutation | AGA,AGG | R,R 200 | XP_011515622.1 | |
XM_017013872.1 | 894 | Silent Mutation | AGA,AGG | R,R 261 | XP_016869361.1 | |
XM_017013873.1 | 894 | Silent Mutation | AGA,AGG | R,R 261 | XP_016869362.1 | |
XM_017013874.1 | 894 | Silent Mutation | AGA,AGG | R,R 261 | XP_016869363.1 | |
XM_017013875.1 | 894 | Silent Mutation | AGA,AGG | R,R 240 | XP_016869364.1 | |
XM_017013876.1 | 894 | Silent Mutation | AGA,AGG | R,R 200 | XP_016869365.1 |