Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCAGTGCAATGAAAAGACTTAA[A/G]ATCATATATATAAAGAGGCTGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607400 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCOLN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCOLN3 - mucolipin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253693.1 | 1384 | Silent Mutation | ATC,ATT | I,I 437 | NP_001240622.1 | |
NM_018298.10 | 1384 | Silent Mutation | ATC,ATT | I,I 493 | NP_060768.8 | |
XM_005271003.1 | 1384 | Silent Mutation | ATC,ATT | I,I 493 | XP_005271060.1 | |
XM_006710750.1 | 1384 | Silent Mutation | ATC,ATT | I,I 493 | XP_006710813.1 | |
XM_011541739.2 | 1384 | Silent Mutation | ATC,ATT | I,I 506 | XP_011540041.2 | |
XM_011541740.2 | 1384 | Silent Mutation | ATC,ATT | I,I 461 | XP_011540042.1 | |
XM_011541741.2 | 1384 | Intron | XP_011540043.1 | |||
XM_011541742.1 | 1384 | Silent Mutation | ATC,ATT | I,I 338 | XP_011540044.1 |