Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAGGGCCGCCGGAGCCAGAGCC[A/G]CAGGAGCCACCGCAGCCGGAGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125265 MIM: 182175 MIM: 601079 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
REEP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
REEP5 - receptor accessory protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005669.4 | 1861 | Intron | NP_005660.4 | |||
XM_017009844.1 | 1861 | Intron | XP_016865333.1 |
SRP19 - signal recognition particle 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204193.1 | 1861 | Intron | NP_001191122.1 | |||
NM_001204194.1 | 1861 | Intron | NP_001191123.1 | |||
NM_001204196.1 | 1861 | Intron | NP_001191125.1 | |||
NM_001204199.1 | 1861 | UTR 3 | NP_001191128.1 | |||
NM_003135.2 | 1861 | Intron | NP_003126.1 |
ZRSR1 - zinc finger CCCH-type, RNA binding motif and serine/arginine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |