Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCTGCGAAGAAGAGGCCTGGCC[A/G]TGGGTGTATGGGGGAAGAAACATCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613902 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929234 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
LOC101929234 - uncharacterized LOC101929234 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF503 - zinc finger protein 503 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF503-AS2 - ZNF503 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |