Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCACATTGTTTATGTGTATGTG[A/G]CTTCTCTCCATATTCCTGACACTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF442 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF442 - zinc finger protein 442 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030824.2 | 493 | Missense Mutation | CCA,TCA | P,S 152 | NP_110451.1 | |
XM_006722908.3 | 493 | Missense Mutation | CCA,TCA | P,S 145 | XP_006722971.1 | |
XM_017027316.1 | 493 | Missense Mutation | CCA,TCA | P,S 160 | XP_016882805.1 | |
XM_017027317.1 | 493 | Missense Mutation | CCA,TCA | P,S 95 | XP_016882806.1 | |
XM_017027318.1 | 493 | Missense Mutation | CCA,TCA | P,S 152 | XP_016882807.1 |
ZNF563 - zinc finger protein 563 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |