Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCGCCACCCTCACCCCCATCAG[T/C]GTTTGGATGCCTTCCTCTAGGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603515 MIM: 139250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSHL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSHL1 - chorionic somatomammotropin hormone like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GH1 - growth hormone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000515.4 | 523 | Silent Mutation | ACA,ACG | T,T 149 | NP_000506.2 | |
NM_022559.3 | 523 | Silent Mutation | ACA,ACG | T,T 134 | NP_072053.1 | |
NM_022560.3 | 523 | Silent Mutation | ACA,ACG | T,T 109 | NP_072054.1 |