Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGGCAGGGGAGGAGGGCAAAAG[G/T]TGTTGAGCCAAGCTGGATGGTTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608198 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGLYRP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGLYRP4 - peptidoglycan recognition protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020393.3 | 1576 | UTR 3 | NP_065126.2 | |||
XM_011509789.2 | 1576 | UTR 3 | XP_011508091.1 | |||
XM_011509790.1 | 1576 | Intron | XP_011508092.1 | |||
XM_011509791.2 | 1576 | UTR 3 | XP_011508093.1 | |||
XM_011509792.1 | 1576 | Intron | XP_011508094.1 | |||
XM_011509793.2 | 1576 | UTR 3 | XP_011508095.1 |