Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGGAAGATGTTGGTAACATGG[A/G]GGTTCAAATATGAGACATAAAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 177046 MIM: 170260 MIM: 170261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMB8 - proteasome subunit beta 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004159.4 | Intron | NP_004150.1 | ||||
NM_148919.3 | Intron | NP_683720.2 |
PSMB8-AS1 - PSMB8 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAP1 - transporter 1, ATP binding cassette subfamily B member | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAP2 - transporter 2, ATP binding cassette subfamily B member | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |